MilliporeSigma
SPR detection of cardiac troponin T for acute myocardial infarction.
Talanta (2015-12-24)
Maria Pawula, Zeynep Altintas, Ibtisam E Tothill
ABSTRACT
A surface plasmon resonance (SPR) sensor developed for the rapid, sensitive and specific detection of cardiac troponin T (cTnT) in serum samples is reported in this work. An extensive optimisation of assay parameters was conducted to achieve optimal detection strategy. Both direct and sandwich immunoassay formats were investigated and optimised. The response obtained was enhanced further by the use of gold nanoparticles (AuNPs) conjugated to the anti-cTnT detection antibody. A regeneration method was developed to enable the reuse of the SPR sensor for multiple sample application. The SPR immunosensor showed good reproducibility for cTnT detection in the concentration range of 25-1000 ng mL(-1) and 5-400 ng mL(-1) for the direct and sandwich assays in buffer, respectively. The linear regression analysis was performed and R(2) value was found as 0.99 for both assays. In order to optimise the sensor for serum analysis, nonspecific binding of serum proteins was reduced through the use of additives in the dilution buffer. To achieve greater sensitivity, the performance of the cTnT immunosensor sandwich assay in human serum was evaluated using non-modified and AuNP modified detector antibodies. A detection limit (LOD) for the immunosensor in 50% serum was assessed as 5 ng mL(-1) cTnT for the standard sandwich assay and 0.5 ng mL(-1) cTnT when using AuNP conjugated detector antibodies with a linear dynamic range of 0.5-40 ng mL(-1). The dissociation constant was found as 3.28 × 10(-9) M using Langmuir binding model which indicates high affinity between cTnT and its antibody. The proposed SPR immunosensor has a promising potential to be developed for point-of-care testing for the early diagnosis of acute myocardial infarction (AMI). This method can also be used for the rapid detection of biomarkers in central nervous system diseases.
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%