  • Search Results
  • Characterization of human ethmoid sinus mucosa derived mesenchymal stem cells (hESMSCs) and the application of hESMSCs cell sheets in bone regeneration.
Characterization of human ethmoid sinus mucosa derived mesenchymal stem cells (hESMSCs) and the application of hESMSCs cell sheets in bone regeneration.
Biomaterials (2015-07-22)
Qing Xie, Zi Wang, Yazhuo Huang, Xiaoping Bi, Huifang Zhou, Ming Lin, Zhang Yu, Yefei Wang, Ni Ni, Jing Sun, Si Wu, Zhengwei You, Chunyu Guo, Hao Sun, Yadong Wang, Ping Gu, Xianqun Fan
ABSTRACT
Mesenchymal stem cells (MSCs) have been extensively applied in the field of tissue regeneration. MSCs derived from various tissues exhibit different characteristics. In this study, a cluster of cells were isolated from human ethmoid sinus mucosa membrane and termed as hESMSCs. hESMSCs was demonstrated to have MSC-specific characteristics of self-renewal and tri-lineage differentiation. In particular, hESMSCs displayed strong osteogenic differentiation potential, and also remarkably promoted the proliferation and osteogenesis of rat bone marrow mesenchymal stem cells (rBMSCs) in vitro. Next, hESMSCs were prepared into a cell sheet and combined with a PSeD scaffold seeded with rBMSCs to repair critical-sized calvarial defects in rats, which showed excellent reparative effects. Additionally, ELISA assays revealed that secreted cytokines, such as BMP-2, BMP-4 and bFGF, were higher in the hESMSCs conditioned medium, and immunohistochemistry validated that hESMSCs cell sheet promoted the expression of BMP signaling downstream genes in newly formed bone. In conclusion, hESMSCs were demonstrated to be a class of mesenchymal stem cells that possessed high self-renewal capacity along with strong osteogenic potential, and the cell sheet of hESMSCs could remarkably promote new bone regeneration, indicating that hESMSCs cell sheet could serve as a novel and promising alternative strategy in the management of bone regeneration.
Sigma-Aldrich
Alizarin, Dye content 97 %
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Sulfobromophthalein disodium salt hydrate, used to study hepatocyte transport functions
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)