  • Search Results
  • Cationic Polyrotaxanes as a Feasible Framework for the Intracellular Delivery and Sustainable Activity of Anionic Enzymes: A Comparison Study with Methacrylate-Based Polycations.
Cationic Polyrotaxanes as a Feasible Framework for the Intracellular Delivery and Sustainable Activity of Anionic Enzymes: A Comparison Study with Methacrylate-Based Polycations.
Macromolecular bioscience (2015-04-30)
Atsushi Tamura, Go Ikeda, Kei Nishida, Nobuhiko Yui
ABSTRACT
We have developed cationic polyrotaxanes composed of N,N-dimethylaminoethyl (DMAE) group-modified α-cyclodextrins (α-CDs) threaded along a poly(ethylene glycol) (PEG) chain capped with a terminal bulky stopper (DMAE-PRX) for the intracellular delivery of proteins through the polyelectrolyte complexation. Herein, to ascertain the effect of supramolecular backbone structure of cationic polyrotaxanes, the physicochemical properties and biological activity of polyelectrolyte complex with anionic β-galactosidase (β-gal) were investigated in comparison to a cationic linear polymer, poly[2-(N,N-dimethylaminoethyl) methacrylate] (PDMAEMA). In the cellular experiments, the DMAE-PRX/β-gal complexes exhibited higher intracellular uptake of β-gal and sustainable enzymatic activity of delivered β-gal than the PDMAEMA/β-gal complexes. It is considered that the cationic polyrotaxanes are promising supramolecular backbone structure for the intracellular protein delivery.
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
2-Nitrophenyl β-D-galactopyranoside, ≥98% (enzymatic)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
2-Cyano-2-propyl benzodithioate, >97% (HPLC)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Isobutyronitrile, 99.6%
Sigma-Aldrich
Isobutyronitrile, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis