  • Search Results
  • Counter-pressure-assisted ITP with electrokinetic injection under field-amplified conditions for bacterial analysis.
Counter-pressure-assisted ITP with electrokinetic injection under field-amplified conditions for bacterial analysis.
Analytical and bioanalytical chemistry (2015-07-06)
Sui Ching Phung, Yi Heng Nai, Mirek Macka, Shane M Powell, Rosanne M Guijt, Michael C Breadmore
ABSTRACT
Counter-pressure was used to extend the duration of field-amplified sample injection in isotachophoresis (FASI-ITP) in order to improve the detection of bacterial cells. Using 0.51-μm negatively charged encapsulated fluorescent beads as a model, the counter-pressure, injection and separation voltages, and times were optimized. Using 6-min 8,963-Pa counter-pressure FASI-ITP injections at -12 kV followed by mobilization of the ITP band with continued injection at -6 kV, the limit of detection (LOD) for Escherichia coli was improved to 78 cells/mL, a factor of 4 when compared with FASI-ITP without counter-pressure.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
SAFC
Tromethamine