  • Search Results
  • Simultaneous analysis of herbicides pendimethalin, oxyfluorfen, imazethapyr and quizalofop-p-ethyl by LC-MS/MS and safety evaluation of their harvest time residues in peanut (Arachis hypogaea L.).
Simultaneous analysis of herbicides pendimethalin, oxyfluorfen, imazethapyr and quizalofop-p-ethyl by LC-MS/MS and safety evaluation of their harvest time residues in peanut (Arachis hypogaea L.).
Journal of food science and technology (2015-07-04)
Ajoy Saha, Ahammed Shabeer T P, Kaushik Banerjee, Sandip Hingmire, Debarati Bhaduri, N K Jain, Sagar Utture
ABSTRACT
This paper reports a simple and rapid method for simultaneous determination of the residues of selected herbicides viz. pendimethalin, oxyfluorfen, imazethapyr and quizalofop-p-ethyl in peanut by liquid chromatography-tandem mass spectrometry (LC-MS/MS). A modified approach of the QuEChERS methodology was used to extract the herbicides from the peanut kernel without any clean-up. The method showed excellent linearity (r(2) > 0.99) with no significant matrix effect. Accuracy of the method in terms of average recoveries of all the four herbicides ranged between 69.4 -94.4 % at spiking levels of 0.05, 0.10 and 0.25 mg kg(-1) with intra-day and inter-day precision RSD (%) between 2.6-16.6 and 8.0-11.3, respectively. Limit of quantification (LOQs) was 5.0 μg kg(-1) for pendimethalin, imazethapyr and quizalofop-p-ethyl and 10.0 μg kg(-1) for oxyfluorfen. The expanded uncertainties were <11 % for determination of these herbicides in peanut. The proposed method was successfully applied for analysis of these herbicide residues in peanut samples harvested from the experimental field and the residues were below the detection level.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium chloride, tablet