Bioactive products formed in humans from fish oils.
Journal of lipid research (2015-07-17)
Carsten Skarke, Naji Alamuddin, John A Lawson, Xuanwen Li, Jane F Ferguson, Muredach P Reilly, Garret A FitzGerald
ABSTRACT
Resolvins, maresins, and protectins can be formed from fish oils. These specialized pro-resolving mediators (SPMs) have been implicated in the resolution of inflammation. Synthetic versions of such SPMs exert anti-inflammatory effects in vitro and when administered to animal models. However, their importance as endogenous products formed in sufficient amounts to exert anti-inflammatory actions in vivo remains speculative. We biased our ability to detect SPMs formed in healthy volunteers by supplementing fish oil in doses shown previously to influence blood pressure and platelet aggregation under placebo-controlled conditions. Additionally, we sought to determine the relative formation of SPMs during an acute inflammatory response and its resolution, evoked in healthy volunteers by bacterial lipopolysaccharide (LPS). Bioactive lipids, enzymatic epoxyeicosatrienoic acids (EETs), and free radical-catalyzed prostanoids [isoprostanes (iPs)] formed from arachidonic acid and the fish oils, served as comparators. Despite the clear shift from ω-6 to ω-3 EETs and iPs, we failed to detect a consistent signal, in most cases, of SPM formation in urine or plasma in response to fish oil, and in all cases in response to LPS on a background of fish oil. Our results question the relevance of these SPMs to the putative anti-inflammatory effects of fish oils in humans.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Dehydro-L-(+)-ascorbic acid dimer, ≥80% (enzymatic)
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C