  • Search Results
  • Cloning, Expression, and Purification of Recombinant Uricase Enzyme from Pseudomonas aeruginosa Ps43 Using Escherichia coli.
Cloning, Expression, and Purification of Recombinant Uricase Enzyme from Pseudomonas aeruginosa Ps43 Using Escherichia coli.
Journal of microbiology and biotechnology (2015-01-16)
Mona I Shaaban, Eman Abdelmegeed, Youssif M Ali
ABSTRACT
Uricase is an important microbial enzyme that can be used in the clinical treatment of gout, hyperuricemia, and tumor lysis syndrome. A total of 127 clinical isolates of Pseudomonas aeruginosa were tested for uricase production. A Pseudomonas strain named Ps43 showed the highest level of native uricase enzyme expression. The open reading frame of the uricase enzyme was amplified from Ps43 and cloned into the expression vector pRSET-B. Uricase was expressed using E. coli BL21 (DE3). The ORF was sequenced and assigned GenBank Accession No. KJ718888. The nucleotide sequence analysis was identical to the coding sequence of uricase gene puuD of P. aeruginosa PAO1. We report the successful expression of P. aeruginosa uricase in Escherichia coli. E. coli showed an induced protein with a molecular mass of about 58 kDa that was confirmed by sodium dodecyl sulfate-polyacrylamide gel electrophoresis and Western blotting. We also established efficient protein purification using the Ni-Sepharose column with activity of the purified enzyme of 2.16 IU and a 2-fold increase in the specific activity of the pure enzyme compared with the crude enzyme.
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside