MilliporeSigma
Structural Features and Asymmetric Environment of i-Pr-SPRIX Ligand.
Chirality (2015-06-23)
Kazuhiro Takenaka, Xianjin Lin, Shinobu Takizawa, Hiroaki Sasai
ABSTRACT
Novel chiral diisopropyl spiro bis(isoxazoline) ligands, anti-i-Pr-SPRIX and syn-i-Pr-SPRIX, were designed and synthesized. Their catalytic utility, X-ray crystallographic analyses, and complexation studies demonstrated the structural features of tetraisopropyl spiro bis(isoxazoline) ligand, i-Pr-SPRIX, which is a prominent ligand in various enantioselective Pd catalytic processes: All i-Pr groups work in collaboration to create an effective asymmetric environment.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%