  • Search Results
  • The effect of sunitinib on the plasma exposure of intravenous paracetamol and its major metabolite: paracetamol glucuronide.
The effect of sunitinib on the plasma exposure of intravenous paracetamol and its major metabolite: paracetamol glucuronide.
European journal of drug metabolism and pharmacokinetics (2014-03-29)
Agnieszka Karbownik, Edyta Szałek, Katarzyna Sobańska, Wojciech Połom, Tomasz Grabowski, Anna Biczysko-Murawa, Marcin Matuszewski, Anna Wolc, Edmund Grześkowiak
ABSTRACT
The study aimed to examine the effect of sunitinib on the plasma exposure of intravenous paracetamol and its major metabolite, paracetamol glucuronide. Both drugs share metabolic pathways in the liver, and the drug interactions between sunitinib and paracetamol administered in higher doses were reported. These interactions resulted in hepatotoxicity. The adult New Zealand male rabbits were divided into three groups (6 animals each): rabbits receiving sunitinib and paracetamol (SUN + PC), rabbits receiving sunitinib (SUN), and a control group receiving paracetamol (PC). Sunitinib was administered orally (25 mg) and paracetamol was administrated intravenously (35 mg/kg). Blood samples for sunitinib and SU12662 assays were collected up to 96 h after drug administration and for paracetamol and paracetamol glucuronide up to 300 min after drug administration. Aspartate aminotransferase (AST), alanine aminotransferase (ALT), and bilirubin were analysed before and after drug administration. A number of pharmacokinetic parameters were analysed. There were no differences in the levels of AST, ALT, and bilirubin among the groups at either time point. Significantly higher values of AUC0-t , AUC0-∞ , and C max and lower clearance and volume of distribution of paracetamol were observed in group PC vs. group SUN + PC (p < 0.01). The maximum plasma concentration of paracetamol glucuronide tended to be higher in group PC 213.27 μg/mL (90 % CI 1.06, 1.25; p = 0.0267). Statistically significant differences were revealed for paracetamol glucuronide mean residence time (MRT); MRT was higher in group SUN + PC than in group PC (p = 0.0375). The mean t max of paracetamol glucuronide was similar in both groups: SUN + PC and group PC (15 and 20 min, respectively). The mean t max of sunitinib was different in groups SUN + PC and SUN (10.0 and 7.0, respectively; p = 0.0134). At the studied doses, neither of the drugs, whether administered alone or together, had hepatotoxic effects. The present study was not able to confirm that sunitinib, administered at low doses in conjunction with paracetamol, displays a hepatoprotective effect. Significant differences were observed in some pharmacokinetic parameters of paracetamol.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
p-Acetamidophenyl β-D-glucuronide sodium salt
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder