  • Search Results
  • New Flavones, a 2-(2-Phenylethyl)-4H-chromen-4-one Derivative, and Anti-Inflammatory Constituents from the Stem Barks of Aquilaria sinensis.
New Flavones, a 2-(2-Phenylethyl)-4H-chromen-4-one Derivative, and Anti-Inflammatory Constituents from the Stem Barks of Aquilaria sinensis.
Molecules (Basel, Switzerland) (2015-11-28)
Sin-Ling Wang, Tsong-Long Hwang, Mei-Ing Chung, Ping-Jyun Sung, Chih-Wen Shu, Ming-Jen Cheng, Jih-Jung Chen
ABSTRACT
In the current study, two new flavones, 4'-O-geranyltricin (1) and 3'-O-geranylpolloin (2), and a new 2-(2-phenylethyl)-4H-chromen-4-one derivative, 7-hydroxyl-6-methoxy-2-(2-phenylethyl)chromone (3), have been isolated from the stem barks of A. sinensis, together with 21 known compounds 4-24. The structures of new compounds 1-3 were determined through spectroscopic and MS analyses. Compounds 2, 3, 5, 6, and 8-10 exhibited inhibition (IC50≤12.51 μM) of superoxide anion generation by human neutrophils in response to formyl-L-methionyl-L-leucyl-L-phenylalanine/cytochalasin B (fMLP/CB). Compounds 3, 6, 8, 10, and 19 inhibited fMLP/CB-induced elastase release with IC50 values≤15.25 μM. This investigation reveals bioactive isolates (especially 2, 3, 5, 6, 8, 9, 10, and 19) could be further developed as potential candidates for the treatment or prevention of various inflammatory diseases.
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Mercury, electronic grade, 99.9999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Mercury, ≥99.99% trace metals basis
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%