  • Search Results
  • The microRNA-325 inhibits hepatocellular carcinoma progression by targeting high mobility group box 1.
The microRNA-325 inhibits hepatocellular carcinoma progression by targeting high mobility group box 1.
Diagnostic pathology (2015-07-22)
Huifen Li, Weihua Huang, Rongcheng Luo
ABSTRACT
MicroRNAs (miRNAs) can serve as tumor suppressors and might provide an efficient strategy for annihilating tumor cells. Nevertheless, the potential role of miR-325 in hepatocellular carcinoma (HCC) is still unknown. Using RT-PCR, immunoblots invasion assays and bioinformatics strategies, we investigated the potential role of miR-325 in HCC. We showed that miR-325 was decreased and HMGB1 was increased in 99 patients with hepatocellular carcinoma. MiR-325 inhibition promoted cell invasion and proliferation, while miR-325 upregulation inhibited cell invasion and proliferation by using transwell and CCK8 assays. We further showed that HMGB1 might be a direct target of miR-325 and is negatively regulated by miR-325. Down-regulation of miR-325 predicts poor prognosis for HCC patients. These findings implied that miR-325 regulates cell invasion and proliferation via targeting HMGB1 and may be a potential prognostic marker for HCC. The virtual slide(s) for this article can be found here: http://www.diagnosticpathology.diagnomx.eu/vs/4655707031717989.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri