  • Search Results
  • Epigenetic Modulation of Microglial Inflammatory Gene Loci in Helminth-Induced Immune Suppression: Implications for Immune Regulation in Neurocysticercosis.
Epigenetic Modulation of Microglial Inflammatory Gene Loci in Helminth-Induced Immune Suppression: Implications for Immune Regulation in Neurocysticercosis.
ASN neuro (2015-07-08)
Arun Chauhan, Fredice Z Quenum, Ata Abbas, David S Bradley, Sergei Nechaev, Brij B Singh, Jyotika Sharma, Bibhuti B Mishra
ABSTRACT
In neurocysticercosis, parasite-induced immune suppressive effects are thought to play an important role in enabling site-specific inhibition of inflammatory responses to infections. It is axiomatic that microglia-mediated (M1 proinflammatory) response causes central nervous system inflammation; however, the mechanisms by which helminth parasites modulate microglia activation remain poorly understood. Here, we show that microglia display a diminished expression of M1-inflammatory mediators such as tumor necrosis factor-alpha (TNF-α), interleukin-6 (IL-6), and nitric oxide synthase 2 (NOS2) in murine neurocysticercosis. Microglia also exhibited a lack of myeloid cell maturation marker major histocompatibility complex (MHC)-II in these parasite-infected brains. Treatment of microglia with helminth soluble/secreted factors (HSFs) in vitro did not induce expression of M1-inflammatory signature molecule NOS2 as well as MHC-II in primary microglia. However, HSF treatment completely inhibited lipopolysaccharide-induced increase in expression of MHC-II, NOS2 and nitric oxide production in these cells. As epigenetic modulation of chromatin states that regulates recruitment of RNA polymerase II (Pol-II) is a key regulatory step in determining gene expression and functional outcome, we next evaluated whether HSF induced modulation of these phenomenon in microglia in vitro. Indeed, HSF downregulated Pol-II recruitment to the promoter region of TNF-α, IL-6, NOS2, MHC-II, and transcription factor CIITA (a regulator of MHC-II expression), by itself. Moreover, HSF suppressed the lipopolysaccharide-induced increase in Pol-II recruitment as well. In addition, HSF exposure reduced the positive histone marks H3K4Me3 and H3K9/14Ac at the promoter of TNF-α, IL-6, NOS2, MHC-II, and CIITA. These studies provide a novel mechanistic insight into helminth-mediated immune suppression in microglia via modulation of epigenetic processes.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine