  • Search Results
  • Evaluation of body growth and immunity-related differentially expressed genes through deep RNA sequencing in the piglets of Jeju native pig and Berkshire.
Evaluation of body growth and immunity-related differentially expressed genes through deep RNA sequencing in the piglets of Jeju native pig and Berkshire.
Animal genetics (2015-03-11)
M Ghosh, S S Sodhi, K-D Song, J H Kim, R K Mongre, N Sharma, N K Singh, S W Kim, H K Lee, D K Jeong
ABSTRACT
This study was carried out with the objective to investigate the differentially expressed genes (DEGs) between Jeju native pig (JNP) and Berkshire piglets. The RNA-Seq technique was used to investigate the transcriptomes in the fat, liver and longissimus dorsi muscle from these two breeds. Paired-end reads of the sequences that passed the quality filters were aligned to the Sus scrofa genome using tophat2 (v2.0.2). In this study, 65% of muscle, 20% of fat and 54% of liver genes showed higher expression in the piglets of JNP than in Berkshire. Gene Ontology and signaling pathways showed that immune response and lipid metabolisms were commonly enriched pathways in all three tissues. It was found that the genes pertaining to body growth and immune system are significantly (P < 0.01) more highly expressed in Berkshire piglets. DEGs explored between the piglets of the two breeds might influence the identification of the genetic markers for further breed improvement programs. Our findings provide a new perspective for understanding and identifying candidate genes that are involved in various biological functions. Moreover, transcriptome analysis makes it easier to understand the differences between genetic mechanisms of breeds.
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%