  • Search Results
  • Human hydroxylated metabolites of BDE-47 and BDE-99 are glucuronidated and sulfated in vitro.
Human hydroxylated metabolites of BDE-47 and BDE-99 are glucuronidated and sulfated in vitro.
Toxicology letters (2015-05-10)
Claudio Erratico, Xiaobo Zheng, Andreas Ryden, Goran Marsh, Walid Maho, Adrian Covaci
ABSTRACT
Polybrominated diphenyl ethers (PBDEs) were used worldwide as additive flame retardants and are classified as persistent, bioaccumulable and toxic environmental pollutants. In humans, the hydroxylated metabolites of 2,2',4,4'-tetrabromodiphenyl ether (BDE-47) and 2,2',4,4',5-pentabromodiphenyl ether (BDE-99) formed in vitro have also been detected in vivo. To further characterize the metabolism of BDE-47 and BDE-99 and to identify candidate markers for monitoring the human exposure to PBDEs using non-invasive approaches, glucuronidation and sulfation of hydroxylated metabolites of BDE-47 and BDE-99 were investigated using human liver microsomes and cytoplasm, respectively. The formed Phase II metabolites were analyzed by liquid chromatography-tandem mass spectrometry using a novel approach to develop analytical methods in absence of authentic standards. All available standards for hydroxylated metabolites of BDE-47 and BDE-99 were glucuronidated and sulfated, showing that glucuronidation and sulfation are part of the metabolism pathway of BDE-47 and BDE-99 in vitro. The major glucuronidated and sulfated analogs of hydroxylated metabolites of BDE-47 were (a) 2,4-DBP-Gluc and 5-Gluc-BDE-47, and (b) 2'-Sulf-BDE-28, 4-Sulf-BDE-42 and 3-Sulf-BDE-47, respectively. The major glucuronidated and sulfated analogs of hydroxylated metabolites of BDE-99 were (a) 2,4,5-TBP-Gluc and 6'-Gluc-BDE-99, and (b) 3'-Sulf-BDE-99 and 5'-Sulf-BDE-99, respectively. Apparent Km values associated with the formation of sulfated metabolites of BDE-47 and BDE-99 were ten times lower than those of the corresponding glucuronidated metabolites, suggesting that sulfated rather than glucuronidated metabolites of OH-PBDEs might be used as markers of human exposure to PBDEs using a non-invasive approach based on urine sample collection.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2,4-Dibromophenol, 95%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)