  • Search Results
  • Mechanistic studies of anticancer aptamer AS1411 reveal a novel role for nucleolin in regulating Rac1 activation.
Mechanistic studies of anticancer aptamer AS1411 reveal a novel role for nucleolin in regulating Rac1 activation.
Molecular oncology (2015-04-26)
E Merit Reyes-Reyes, Francesca R Šalipur, Mitra Shams, Matthew K Forsthoefel, Paula J Bates
ABSTRACT
AS1411 is a G-rich quadruplex-forming oligodeoxynucleotide that binds specifically to nucleolin, a protein found on the surface and in the cytoplasm of most malignant cells but absent from the surface/cytoplasm of most normal cells. AS1411 has shown promising clinical activity and is being widely used as a tumor-targeting agent, but its mechanism of action is not fully understood. Previously, we showed that AS1411 is taken up in cancer cells by macropinocytosis (fluid phase endocytosis) and subsequently stimulates further macropinocytosis by a nucleolin-dependent mechanism. In the current study, we have investigated the significance and molecular mechanisms of AS1411-induced macropinocytosis. Our results indicate that the antiproliferative activity of AS1411 in various cell lines correlated with its capacity to stimulate macropinocytosis. In DU145 prostate cancer cells, AS1411 induced activation of EGFR, Akt, p38, and Rac1. Activation of Akt and p38 were not critical for AS1411 activity because Akt activation was not observed in all AS1411-responsive cell lines and knockdown of p38 had no effect on AS1411's ability to inhibit proliferation. On the other hand, activation of EGFR and Rac1 appeared to play a role in AS1411 activity in all cancer cell lines examined (DU145, MDA-MB-468, A549, LNCaP) and their inhibition significantly reduced AS1411-mediated macropinocytosis and AS1411 antiproliferative activity. Interestingly, downregulation of nucleolin expression by siRNA also produced a substantial increase in activated Rac1, revealing a previously unknown role for nucleolin as a negative regulator of Rac1 activation. Our results are consistent with a model whereby AS1411 binding to nucleolin leads to sustained activation of Rac1 and causes methuosis, a novel type of nonapoptotic cell death characterized by hyperstimulation of macropinocytosis. We speculate that methuosis is a tumor/metastasis suppressor mechanism that opposes the malignant functions of Rac1 and that cancer cells may overexpress nucleolin to surmount this barrier.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ras Activation Assay Kit
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%