  • Search Results
  • Genetic Structure of Pacific Trout at the Extreme Southern End of Their Native Range.
Genetic Structure of Pacific Trout at the Extreme Southern End of Their Native Range.
PloS one (2015-10-29)
Alicia Abadía-Cardoso, John Carlos Garza, Richard L Mayden, Francisco Javier García de León
ABSTRACT
Salmonid fishes are cold water piscivores with a native distribution spanning nearly the entire temperate and subarctic northern hemisphere. Trout in the genus Oncorhynchus are the most widespread salmonid fishes and are among the most important fish species in the world, due to their extensive use in aquaculture and valuable fisheries. Trout that inhabit northwestern Mexico are the southernmost native salmonid populations in the world, and the least studied in North America. They are unfortunately also facing threats to their continued existence. Previous work has described one endemic species, the Mexican golden trout (O. chrysogaster), and one endemic subspecies, Nelson's trout (O. mykiss nelsoni), in Mexico, but previous work indicated that there is vastly more biodiversity in this group than formally described. Here we conducted a comprehensive genetic analysis of this important group of fishes using novel genetic markers and techniques to elucidate the biodiversity of trout inhabiting northwestern Mexico, examine genetic population structure of Mexican trout and their relationships to other species of Pacific trout, and measure introgression from non-native hatchery rainbow trout. We confirmed substantial genetic diversity and extremely strong genetic differentiation present in the Mexican trout complex, not only between basins but also between some locations within basins, with at least four species-level taxa present. We also revealed significant divergence between Mexican trout and other trout species and found that introgression from non-native rainbow trout is present but limited, and that the genetic integrity of native trout is still maintained in most locations. This information will help to guide effective conservation strategies for this important group of fishes.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%