  • Search Results
  • Metabolic suppression during mesodermal differentiation of embryonic stem cells identified by single-cell comprehensive gene expression analysis.
Metabolic suppression during mesodermal differentiation of embryonic stem cells identified by single-cell comprehensive gene expression analysis.
Molecular bioSystems (2015-07-28)
Yuanshu Zhou, Ikuma Fujisawa, Kosuke Ino, Tomokazu Matsue, Hitoshi Shiku
ABSTRACT
Flk-1 (VEGF receptor 2) is a well-defined mesodermal progenitor marker and the Flk-1-positive (Flk-1(+)) cells derived from embryonic stem cells (ESCs) have been known to generate hemangioblasts and cardiovascular progenitor cells, which are formed in the early and late stages of differentiation, respectively. In this study, we separated Flk-1(+) and Flk-1(-) cells from spontaneously differentiating embryoid bodies (EBs) of mouse ESCs. We found that cell aggregates derived from late stage Flk-1(+) cells had a relatively small size and a low oxygen consumption rate (OCR) compared with those derived from Flk-1(-) cells. Furthermore, using single-cell comprehensive gene expression analysis, we found that both Flk-1(+) and Flk-1(-) cells could be categorized into subgroups with either low or high glucose metabolic activity. We observed that metabolic suppression occurs in cells expressing an intermediate level of both Nanog and Pou5f1. Taken together, our data suggested that the temporary metabolic suppression is an intrinsic feature of mesodermal differentiation.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine