  • Search Results
  • An NIR-triggered and thermally responsive drug delivery platform through DNA/copper sulfide gates.
An NIR-triggered and thermally responsive drug delivery platform through DNA/copper sulfide gates.
Nanoscale (2015-07-07)
Lei Zhang, Yecheng Li, Zexun Jin, Jimmy C Yu, King Ming Chan
ABSTRACT
Nanomaterials for effective drug delivery require zero pre-release and on-demand release of therapeutic drugs. In this work we demonstrate a novel drug delivery system composed of a mesoporous silica platform conjugated to CuS nanoparticles with two complementary DNA sequences. CuS nanoparticles act as both gatekeepers preventing pre-release of drugs and photothermal agents for effective killing of cancer cells. This system exhibits temperature and NIR-responsive DOX release, with an additional accelerated release rate with GSH treatment. Therefore, it can act as an effective anticancer drug delivery carrier with triggered drug release and efficient anti-cancer effect in vitro after NIR irradiation.
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Thioacetamide, reagent grade, 98%
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Thioglycolic acid, ≥99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis