  • Search Results
  • Effects of coarse chalk dust particles (2.5-10 μm) on respiratory burst and oxidative stress in alveolar macrophages.
Effects of coarse chalk dust particles (2.5-10 μm) on respiratory burst and oxidative stress in alveolar macrophages.
Environmental science and pollution research international (2015-04-24)
Yuexia Zhang, Zhenhua Yang, Yan Feng, Ruijin Li, Quanxi Zhang, Hong Geng, Chuan Dong
ABSTRACT
The main aim of the present study was to examine in vitro responses of rat alveolar macrophages (AMs) exposed to coarse chalk dust particles (particulate matter in the size range 2.5-10 μm, PM(coarse)) by respiratory burst and oxidative stress. Chalk PM(coarse)-induced respiratory burst in AMs was measured by using a luminol-dependent chemiluminescence (CL) method. Also, the cell viability; lactate dehydrogenase (LDH) release; levels of cellular superoxide dismutase (SOD), catalase (CAT), glutathione (GSH), malondialdehyde (MDA), and acid phosphatase (ACP); plasma membrane ATPase; and extracellular nitric oxide (NO) level were determined 4 h following the treatment with the different dosages of chalk PM(coarse). The results showed that chalk PM(coarse) initiated the respiratory burst of AMs as indicated by strong CL, which was inhibited by diphenyleneiodonium chloride and L-N-nitro-L-arginine methyl ester hydrochloride. It suggested that chalk PM(coarse) induced the production of reactive oxygen species (ROS) and reactive nitrogen species (RNS) in AMs. This hypothesis was confirmed by the fact that chalk PM(coarse) resulted in a significant decrease of intracellular SOD, GSH, ACP, and ATPase levels and a notable increase of intracellular CAT, MDA content, and extracellular NO level, consequently leading to a decrease of the cell viability and a increase of LDH release. It was concluded that AMs exposed to chalk PM(coarse) can suffer from cytotoxicity which may be mediated by generation of excessive ROS/RNS. Graphical Abstract The possible mechanism of coarse chalk particles-induced adverse effects in AMs.
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Nitric oxide, 98.5%
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)