  • Search Results
  • Assessment of the impact of manufacturing changes on the physicochemical properties and biological activity of Her1-ECD vaccine during product development.
Assessment of the impact of manufacturing changes on the physicochemical properties and biological activity of Her1-ECD vaccine during product development.
Vaccine (2015-05-25)
Katia Garcia Duardo, Yadira Prieto Curbelo, Judith Raymond Pous, Estela Yamilet Rabasa Legón, Belinda Sánchez Ramírez, Kathya Rashida de la Luz Hernández, Adolfo Castillo Vitoch
ABSTRACT
Vaccine preparations based on the extracellular domain of Her1 (Her1-ECD) have demonstrated, in vitro and in vivo, a potent antimetastatic effect on EGFR(+) Lewis lung carcinoma model, while associated side effects were absent. The Her1-ECD is a glycoprotein with a molecular weight of 105 kDa and has 11 potential sites for N-glycosylation. Currently Her1-ECD based vaccine has been evaluated in patients with hormone refractory prostate cancer. Her1-ECD molecule used for in clinical trials was obtained from culture supernatant of HEK 293 transfectomes used the protein free culture media and is purified by immunoaffinity chromatography. In order to increase the cell growth and productivity, new defined culture media have been developed (alternative culture media) in Her1-ECD vaccine production process. In this work, a comparability study was performed to evaluate the impact of process changes in the characteristics physic-chemical and biologicals of the Her1-ECD protein and the degree of similitude between both variants. Techniques such as: SDS-PAGE, SEC-HPLC, isoelectric point, peptide mapping, mass spectrometric, SCX-HPLC, oligosaccharide map, ELISA and flow cytometric were used with this aim. Results indicated that this process change decreases the degree of sialylation of the protein but does not affect its biological activity (measured as titers of Abs and recognition for A431 cell line).
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Anthranilamide, ≥98%
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)