MilliporeSigma
A nonapoptotic role for BAX and BAK in eicosanoid metabolism.
ACS chemical biology (2015-03-31)
Tejia Zhang, Loren D Walensky, Alan Saghatelian
ABSTRACT
BCL-2 proteins are key regulators of programmed cell death. The interplay between pro and antiapoptotic BCL-2 members has important roles in many cancers. In addition to their apoptotic function, recent evidence supports key nonapoptotic roles for several BCL-2 proteins. We used an unbiased lipidomics strategy to reveal that the proapoptotic proteins BAX, and to a lesser extent BAK, regulate the cellular inflammatory response by mediating COX-2 expression and prostaglandin biosynthesis. COX-2 upregulation in response to the bacterial endotoxin lipopolysaccharide is blunted in the absence of BAX, and Bax(-/-) mouse embryonic fibroblasts display altered kinetics of NFκB and MAPK signaling following endotoxin treatment. Our approach uncovers a novel, nonapoptotic function for BAX in regulation of the cellular inflammatory response and suggests that inflammation and apoptosis are more tightly connected than previously anticipated.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade