  • Search Results
  • Extraction and characterisation of pomace pectin from gold kiwifruit (Actinidia chinensis).
Extraction and characterisation of pomace pectin from gold kiwifruit (Actinidia chinensis).
Food chemistry (2015-05-16)
Oni Yuliarti, Kelvin K T Goh, Lara Matia-Merino, John Mawson, Charles Brennan
ABSTRACT
Gold kiwifruit pomace extracted using citric acid, water and enzyme (Celluclast 1.5L) were studied in terms of pectin yield, protein, ash, non-starch polysaccharide, galacturonic acid (GalA), neutral sugar composition, molar mass (Mw), viscosity and degree of branching. Water-extracted pectin was considered closest to its native form. Enzyme extracted pectin showed the highest yield (∼ 4.5%w/w) as compared with the acid and water extraction methods (∼ 3.6-3.8%w/w). Pectin obtained from different extraction methods showed different degree of branching. The Mw and root mean square (RMS) radius varied with the extraction methods with values of 8.4 × 10(5) g/mol and 92 nm, 8.5 × 10(5)g/mol and 102 nm, 6.7 × 10(5) g/mol and 52 nm for acid, water and enzymatic extraction methods, respectively. Similar trend was observed for pectin viscosity, with water-extracted pectin giving a slightly higher viscosity followed by acid and enzyme-extracted pectin. This study showed that gold kiwifruit pomace pectin has potential application in food products.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O