  • Search Results
  • The dependency of compound biological effectiveness factors on the type and the concentration of administered neutron capture agents in boron neutron capture therapy.
The dependency of compound biological effectiveness factors on the type and the concentration of administered neutron capture agents in boron neutron capture therapy.
SpringerPlus (2015-02-13)
Shin-Ichiro Masunaga, Yoshinori Sakurai, Hiroki Tanaka, Keizo Tano, Minoru Suzuki, Natsuko Kondo, Masaru Narabayashi, Yosuke Nakagawa, Tsubasa Watanabe, Akira Maruhashi, Koji Ono
ABSTRACT
To examine the effect of the type and the concentration of neutron capture agents on the values of compound biological effectiveness (CBE) in boron neutron capture therapy. After the subcutaneous administration of a (10) B-carrier, boronophenylalanine- (10) B (BPA) or sodium mercaptododecaborate- (10) B (BSH), at 3 separate concentrations, the (10) B concentrations in tumors were measured by γ-ray spectrometry. SCC VII tumor-bearing C3H/He mice received 5-bromo-2'-deoxyuridine (BrdU) continuously to label all intratumor proliferating (P) cells, then treated with BPA or BSH. Immediately after reactor neutron beam irradiation, during which intratumor (10) B concentrations were kept at levels similar to each other, cells from some tumors were isolated and incubated with a cytokinesis blocker. The responses of BrdU-unlabeled quiescent (Q) and total (= P + Q) tumor cells were assessed based on the frequencies of micronucleation using immunofluorescence staining for BrdU. The CBE values were higher in Q cells and in the use of BPA than total cells and BSH, respectively. In addition, the higher the administered concentrations were, the smaller the CBE values became, with a clearer tendency in the use of BPA than BSH. The values for neutron capture agents that deliver into solid tumors more dependently on uptake capacity of tumor cells became more changeable. Tumor characteristics, such as micro-environmental heterogeneity, stochastic genetic or epigenetic changes, or hierarchical organization of tumor cells, are thought to partially influence on the value of CBE, meaning that the CBE value itself may be one of the indices showing the degree of tumor heterogeneity.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)