  • Search Results
  • DNA biosensor-based on fluorescence detection of E. coli O157:H7 by Au@Ag nanorods.
DNA biosensor-based on fluorescence detection of E. coli O157:H7 by Au@Ag nanorods.
Biosensors & bioelectronics (2015-04-02)
Jiadi Sun, Jian Ji, Yanqing Sun, Mandour H Abdalhai, Yinzhi Zhang, Xiulan Sun
ABSTRACT
A novel DNA sensor for the detection of the Escherichia coli O157:H7 (E. coli O157:H7) eaeA gene was constructed using surface enhanced fluorescence (SEF). The spacing distance dependence nature of Au@Ag nanorods surface enhanced fluorescence was investigated when the cy3-labled single strand DNA(ssDNA) and the stem-loop DNA probe modified on the nanorods was co-hybridized. The result revealed that the fluorescence intensity reached the maximum value with the spacing distance of about 10nm between cy3 and the Au@Ag nanorods surface. Based on this result, a fluorescence "ON/OFF" switch for detecting the eaeA gene of E. coli O157:H7 was constructed. Under optimal conditions, the DNA sensor produced a linear range from 10(-17) to 10(-11) M with a correlation coefficient of 0.9947 and a detection limit of 3.33×10(-18) M, and was also found to be specific in targeting eaeA. The DNA sensor demonstrated a new strategy of combining eaeA recognition and Au@Ag nanorods for fluorescence signal enhancement, and increased sensitivity in the detection of bacterial specific genes.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
5-Bromosalicylic acid, technical grade, 90%
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
Sodium salicylate, meets USP testing specifications
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium salicylate, puriss. p.a., ≥99.5% (NT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%