  • Search Results
  • Comparison of orthogonal chromatographic and lectin-affinity microarray methods for glycan profiling of a therapeutic monoclonal antibody.
Comparison of orthogonal chromatographic and lectin-affinity microarray methods for glycan profiling of a therapeutic monoclonal antibody.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-06-27)
Matthew C Cook, Sherif J Kaldas, Gauri Muradia, Michael Rosu-Myles, Jeremy P Kunkel
ABSTRACT
The N-linked glycosylation of four lots of a marketed human therapeutic monoclonal antibody (mAb) was assessed by three orthogonal chromatographic methods and a commercial lectin microarray. For chromatography, the N-glycans were removed enzymatically from the mAbs using PNGase F. Native glycans were determined by HPAEC-PAD using a panel of 21 N-glycan standards and a multi-stage linear gradient eluent profile for sequential analyses of typical neutral and sialylated glycans in one chromatographic run. The monosaccharide contents of these glycans following acid hydrolysis were confirmed by HPAEC-PAD with monosaccharide standards. Glycosylation analysis by HILIC-FD after stoichiometric labelling with two different fluorescent tags (2-AA and 2-AB) enabled direct quantitation. The 2-AA- and 2-AB-labelled versions of the same glycan standard panel yielded distinctive separation profiles suitable for orthogonal identification of mAb glycans. Glycan profiling with the lectin microarray required partial denaturation of the intact mAbs to expose the sequestered Fc N-glycans. Glycosylation fingerprints were obtained using a fluorescently labelled antibody directed against human IgG Fc. Fluorescence intensities from the fingerprints were deconvoluted with a proprietary algorithm to obtain semi-quantitative "glycan structural class" information. Glycosylation analyses of the four mAb lots by these four methods, which separate and detect oligosaccharides according to different principles, provided complementary and corroboratory qualitative and quantitative information. The predominant N-linked structures were core-fucosylated asialo diantennary structures with varying galactosylation. There were also trace amounts of afucosyl and bisected glycans, but no detectable sialylation by any of the four methods. The therapeutic mAb demonstrated a high degree of consistency in the types and amounts of N-linked glycans in the four lots (<6% CV), and between all four analysis methods (<6% CV). The described methods are co-supported by the excellent quantitative agreement of their results, which is particularly notable considering the orthogonality of their separation and detection mechanisms.
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate, 96%