MilliporeSigma
  • Alcohol and cigarette smoke components activate human pancreatic stellate cells: implications for the progression of chronic pancreatitis.
Alcohol and cigarette smoke components activate human pancreatic stellate cells: implications for the progression of chronic pancreatitis.
Alcoholism, clinical and experimental research (2015-10-16)
Alexandra T K Lee, Zhihong Xu, Srinivasa P Pothula, Mishaal B Patel, Romano C Pirola, Jeremy S Wilson, Minoti V Apte
ABSTRACT
Chronic pancreatitis, a known complication of alcohol abuse, is characterized histopathologically by prominent fibrosis. Pancreatic stellate cells (PSCs) are responsible for producing this fibrous tissue in chronic pancreatitis and are activated by alcohol. Progression of alcoholic chronic pancreatitis (as assessed by calcification and fibrosis) is thought to be facilitated by concurrent smoking, but the mechanisms are unknown. This study aimed to (a) determine whether human PSCs (hPSCs) and rat PSCs express nicotinic acetylcholine receptors (nAChRs), which are known to bind 2 important components of cigarette smoke, namely nicotine and nicotine-derived nitrosamine ketone (NNK), and (b) examine the effects of cigarette smoke components in the presence and absence of alcohol on PSC activation in vitro. Western blotting was used to detect the presence of nAChRs in primary cultures of PSCs. Clinically relevant concentrations of cigarette smoke components (either cigarette smoke extract [CSE], NNK, or nicotine) ± ethanol (EtOH) were used to treat primary cultures of PSCs, and stellate cell activation was assessed by cell migration, proliferation, collagen production, and apoptosis. We demonstrate, for the first time, that PSCs express nAChRs (isoforms α3, α7, β, ε) and that the expression of the α7 isoform in hPSCs is induced by CSE + EtOH. We also provide novel findings that PSCs are activated by CSE and NNK (both alone and in combination with EtOH) as evidenced by an increase in cell migration and/or proliferation. Further, we demonstrate that activation of PSCs by CSE + EtOH and NNK + EtOH may be mediated via nAChRs on the cells. PSCs are activated by clinically relevant concentrations of cigarette smoke components (CSE and NNK), alone and in combination with EtOH. Thus, in alcoholics who smoke, progression of pancreatic fibrosis may be facilitated by the combined effects of alcohol and cigarette smoke components on hPSC behavior.
Product Description
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology