  • Search Results
  • Effect of tamoxifen on fatty degeneration and atrophy of rotator cuff muscles in chronic rotator cuff tear: An animal model study.
Effect of tamoxifen on fatty degeneration and atrophy of rotator cuff muscles in chronic rotator cuff tear: An animal model study.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2015-07-01)
Edward Cho, Yue Zhang, Anne Pruznak, H Mike Kim
ABSTRACT
Fatty degeneration of the rotator cuff muscles is an irreversible change resulting from chronic rotator cuff tear and is associated with poor clinical outcomes following rotator cuff repair. We evaluated the effect of Tamoxifen, a competitive estrogen receptor inhibitor, on fatty degeneration using a mouse model for chronic rotator cuff tear. Sixteen adult mice were divided into two diet groups (Tamoxifen vs. Regular) and subjected to surgical creation of a large rotator cuff tear and suprascapular nerve transection in their left shoulder with the right shoulder serving as a control. The rotator cuff muscles were harvested at 16 weeks and subjected to histology and RT-PCR for adipogenic and myogenic markers. Histology showed substantially decreased atrophy and endomysial inflammation in Tamoxifen group, but no significant differences in the amount of intramuscular adipocytes and lipid droplets compared to the Regular group. With RT-PCR, the operated shoulders showed significant upregulation of myogenin and PPAR-γ, and downregulation of myostatin compared to the nonsurgical shoulder. No significant differences of gene expression were found between the two diet groups. Our study demonstrated that tamoxifen diet leads to decreased muscle atrophy and inflammatory changes following chronic rotator cuff tear, but has no apparent effect on adipogenesis.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)