  • Search Results
  • Driving Human Granulosa-Luteal Cells Recovered From In Vitro Fertilization Cycles Toward the Follicular Phase Phenotype.
Driving Human Granulosa-Luteal Cells Recovered From In Vitro Fertilization Cycles Toward the Follicular Phase Phenotype.
Reproductive sciences (Thousand Oaks, Calif.) (2015-02-24)
Alessandra Aparecida Vireque, Jacira Ribeiro Campos, Daniel Blasioli Dentillo, Marcelo Picinin Bernuci, Carolina Oliveira Campos, Marcos Felipe Silva-de-Sá, Rui Alberto Ferriani, Altacílio Aparecido Nunes, Ana Carolina Japur de Sá Rosa-e-Silva
ABSTRACT
Culture systems are available for human granulosa cells (GCs) that perpetuate luteinization. The present study examines the plating density effects and long-term serum-free culture on the in vitro dynamics differentiation of luteinizing human GCs. Cells were cultured in serum-free α-minimum essential medium (α-MEM) or serum-based tissue culture medium (TCM). The time course of GCs morphology and secretion of estradiol (E2), progesterone (P4), and relaxin were analyzed after 48, 96, and 144 hours of culture. Other functional markers as follicle-stimulating hormone/luteinizing hormone receptors and steroidogenic enzymes were investigated at the end of culture. The morphology of an α-MEM cell rather than a TCM cell resembles more closely that seen in vivo. Compared to TCM cultures, α-MEM cells secreted 93.7% and 87.2% more E2 and approximately 7% and 17% of the amount of P4 when cultured at densities of 2 × 10(4) or 4 × 10(4) cells/well, respectively. Relaxin secretion was significantly reduced in α-MEM cultures. α-MEM cells were estrogenic and expressed the CYP19 gene. Levels of CYP17 increased about 8-fold in α-MEM cells above the levels found in TCM cells. Our results reveal new insights into human GCs differentiation in vitro and demonstrate the critical importance of the culture system and cell-plating density on the establishment of estrogenic or progestogenic GC phenotypes.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading