  • Search Results
  • Product and process understanding to relate the effect of freezing method on glycation and aggregation of lyophilized monoclonal antibody formulations.
Product and process understanding to relate the effect of freezing method on glycation and aggregation of lyophilized monoclonal antibody formulations.
International journal of pharmaceutics (2015-04-04)
David Awotwe-Otoo, Cyrus Agarabi, Erik K Read, Scott Lute, Kurt A Brorson, Mansoor A Khan
ABSTRACT
The objective of the study was to analyze the effect of controlled and uncontrolled freezing step of a lyophilization process on the extent of non-enzymatic glycation and aggregation of an IgG1 formulation at two concentrations (1mg/ml and 20mg/ml). The degree of glycation (%) was determined through boronate affinity chromatography and its effect on the formation of soluble aggregates and higher molecular weight species was studied using dynamic light scattering (DLS) and size exclusion chromatography with multi-angle light scattering (SEC-MALS). The effect of non-enzymatic glycation on the secondary structure of the formulations was also studied using circular dichroism (CD) spectroscopy and Fourier transform infrared (FT-IR) spectroscopy. Results indicated that controlled nucleation yielded higher residual moisture contents and significantly lower specific surface areas for the two monoclonal antibody concentrations compared with uncontrolled nucleation cycle (p<0.05). For the two concentrations, uncontrolled nucleation resulted in significantly higher levels of glycation compared with controlled nucleation samples (p<0.05). Further, it was observed that higher storage temperatures (25°C/60% RH) versus 5°C resulted in higher glycation. Even though SEC-MALS analyses of the low concentrated formulations did not reveal the formation of higher molecular weight species, DLS analyses at two storage conditions revealed increases in the hydrodynamic radii and polydispersity index of the reconstituted formulations, suggesting the onset of formation of smaller species in the formulations. CD spectroscopy did not reveal any differences in the secondary structure of the mAb for the two concentrations after lyophilization. In conclusion, the freezing step method impacted the extent of glycation in lyophilized samples and the hydrolyzed component of sucrose was critical for increasing glycation. Even though some level of glycation was observed in lyophilized samples, the native structure of the protein was not affected. Further, it was demonstrated that storage of both lyophilized cakes and reconstituted formulations at higher temperatures could increase the extent of glycation in monoclonal antibody formulations.
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, NMR reference standard