  • Search Results
  • Analytical method for simultaneous determination of bulk and intramolecular (13) C-isotope compositions of acetic acid.
Analytical method for simultaneous determination of bulk and intramolecular (13) C-isotope compositions of acetic acid.
Rapid communications in mass spectrometry : RCM (2015-11-14)
Tarin Nimmanwudipong, Alexis Gilbert, Keita Yamada, Naohiro Yoshida
ABSTRACT
Headspace solid-phase microextraction (HS-SPME) combined with gas chromatography/pyrolysis-gas chromatography/combustion-isotope ratio mass spectrometry (GC/Py-GC/C-IRMS) was developed for the simultaneous determination of the intramolecular and molecular carbon-isotopic composition (δ(13) C value) of acetic acid. The δ(13) C values of carboxyl and methyl carbon were standardized using calibration curves constructed from the regression between the measured δ(13) C values and the δ(13) C values of working standards determined in a previous study. We applied this developed HS-SPME-GC/Py-GC/C-IRMS technique to commercial vinegars. In one injection analysis, the bulk and intramolecular δ(13) C values of pure acetic acid standards can be obtained. The repeatability (1σ) of the bulk δ(13) C values is within ±0.4‰, and that of the δ(13) Ccarboxyl and δ(13) Cmethyl values is within ±0.6‰. The intramolecular δ(13) C values of acetic acid in vinegars exhibit a similar pattern. The average Δδ value (δ(13) CCOOH - δ(13) CCH3 ) is 4.3 ± 2.0‰. The approach presented herein for the molecular and intramolecular δ(13) C determination of acetic acid avoids switching between configuration systems and thereby reduces systematic errors. It is expected to be useful for examining isotope fractionation associated with processes related to organic acid (bio)transformations.
Sigma-Aldrich
(±)-L-Alliin, ≥90% (HPLC)
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm (PDMS/DVB), for use with manual holder, needle size 23 ga
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 60 μm (PDMS/DVB), for use with autosampler/HPLC, needle size 24 ga, StableFlex fiber
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), metal alloy fiber, for use with manual holder
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 24 ga, StableFlex, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME LC Tips, C18, tray of 96 ea
Supelco
IonSense® PDMS/DVB SPE-it Tips, pkg of 96 ea
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME StableFlex fiber assortment kit, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, pkg of 3 ea, fiber L 1 cm