  • Search Results
  • NS5ATP9 Promotes Beclin 1-Dependent Starvation-Induced Autophagy of Hepatoblastoma Cells.
NS5ATP9 Promotes Beclin 1-Dependent Starvation-Induced Autophagy of Hepatoblastoma Cells.
Journal of cellular biochemistry (2015-02-05)
Min Quan, Shunai Liu, Qi Wang, Guoli Li, Yu Zhang, Shenghu Feng, Jinqiu Liang, Jun Cheng
ABSTRACT
NS5ATP9, a gene up-regulated by NS5A, plays a crucial oncogenic role in several types of human tumours. However, the underlying mechanisms remain unclear. Autophagy, an evolutionarily conserved catabolic process, maintains cellular homeostasis under stress conditions, such as starvation, and plays a crucial role in tumour initiation and progression. Here, we report that NS5ATP9 mRNA and protein expression was up-regulated in starved HepG2 cells and that the up-regulated NS5ATP9 played a functional role in starvation-induced autophagy. Overexpression or silencing of this gene showed contrasting effects on Beclin 1 and on starvation-induced autophagy. Furthermore, NS5ATP9-mediated autophagy is required for promotion of tumour cell growth, and this effect could be inhibited with 3-methyladenine, chloroquine or by Beclin 1-silencing. Thus, the mechanism for NS5ATP9-promoted autophagy is Beclin 1-dependent in the condition of starvation, and for hepatoblastoma cell growth is also Beclin 1-dependent.
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
BIS-TRIS
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
BIS-TRIS
Sigma-Aldrich
DAPI, for nucleic acid staining