  • Search Results
  • Comparison of thyroid hormone-dependent gene responses in vivo and in organ culture of the American bullfrog (Rana (Lithobates) catesbeiana) lung.
Comparison of thyroid hormone-dependent gene responses in vivo and in organ culture of the American bullfrog (Rana (Lithobates) catesbeiana) lung.
Comparative biochemistry and physiology. Part D, Genomics & proteomics (2015-10-16)
Nik Veldhoen, Mitchel R Stevenson, Caren C Helbing
ABSTRACT
Postembryonic frog development requires a thyroid hormone (TH)-dependent metamorphic transition from an aquatic larva to a terrestrial frog. Such change in environment involves lung maturation in preparation for breathing air. However, little is known regarding the underlying molecular events and the role of THs in this process. Using quantitative real-time polymerase chain reaction, we evaluated Rana (Lithobates) catesbeiana lung mRNA transcripts representing key elements of TH and oxidative stress signaling pathways during natural and TH-induced precocious metamorphosis. TH induction was evaluated in two ways: 1) in vivo through interperitoneal injection of 10pmol/g body weight of 3,3', 5-triiodothyronine (T3) into premetamorphic tadpoles and analysis after 48h, and 2) in serum-free organ culture in the presence of 10nM T3 after 48h. Abundance of transcripts encoding the transcriptional regulators TH receptors α and β, TH-induced bZip protein, and CCAAT/enhancer binding protein 1 was increased during postembryonic development and following administration of exogenous THs to premetamorphic tadpoles in vivo and culture. In contrast, mRNA representing Krüppel-like factor 9 and cold-inducible RNA binding protein revealed differential effects between natural and precocious metamorphosis. Elevated levels of catalase and Cu/Zn superoxide dismutase mRNA were observed at the end of metamorphosis with transcript levels displaying minimal TH-dependency. No change in stress-responsive heat shock protein 30 mRNA abundance was noted. The results support a role for TH-dependent reprogramming of the lung transcriptome during frog development and reveal a requirement for increased antioxidant capacity following anuran metamorphosis.
Sigma-Aldrich
Glycerol solution, puriss. p.a., 86-89% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine