  • Search Results
  • 3,4-Dihydroxy-benzohydroxamic acid (Didox) suppresses pro-inflammatory profiles and oxidative stress in TLR4-activated RAW264.7 murine macrophages.
3,4-Dihydroxy-benzohydroxamic acid (Didox) suppresses pro-inflammatory profiles and oxidative stress in TLR4-activated RAW264.7 murine macrophages.
Chemico-biological interactions (2015-04-07)
Thabe M Matsebatlela, Amy L Anderson, Vincent S Gallicchio, Howard Elford, Charles D Rice
ABSTRACT
Didox (3,4-dihydroxy-benzohydroxamic acid), is a synthetic ribonucleotide reductase (RR) inhibitor derived from polyhydroxy-substituted benzohydroxamic acid, and originally developed as an anti-cancer agent. Some studies indicate that didox may have anti-oxidative stress-like properties, while other studies hint that didox may have anti-inflammatory properties. Using nitric oxide production in response to LPS treatment as a sensitive screening assay for anti-inflammatory compounds, we show that didox is very potent at levels as low as 6.25 μM, with maximal inhibition at 100 μM. A qRT-PCR array was then employed to screen didox for other potential anti-inflammatory and anti-oxidative stress-related properties. Didox was very potent in suppressing the expression of these arrayed mRNA in response to LPS, and in some cases didox alone suppressed expression. Using qRT-PCR as a follow up to the array, we demonstrated that didox suppresses LPS-induced mRNA levels of iNOS, IL-6, IL-1, TNF-α, NF-κβ (p65), and p38-α, after 24h of treatment. Treatment with didox also suppresses the secretion of nitric oxide, IL-6, and IL-10. Furthermore, oxidative stress, as quantified by intracellular ROS levels in response to macrophage activators LPS and phorbol ester (PMA), and the glutathione depleting agent BSO, is reduced by treatment with didox. Moreover, we demonstrate that nuclear translocation of NF-κβ (p65) in response to LPS is inhibited by didox. These findings were supported by qRT-PCR for oxidative stress genes SOD1 and catalase. Overall, this study supports the conclusion that didox may have a future role in managing acute and chronic inflammatory diseases and oxidative stress due to high production of ROS.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture