  • Search Results
  • Polymer Stabilized Fe3O4-Graphene as an Amphiphilic Drug Carrier for Thermo-Chemotherapy of Cancer.
Polymer Stabilized Fe3O4-Graphene as an Amphiphilic Drug Carrier for Thermo-Chemotherapy of Cancer.
ACS applied materials & interfaces (2015-03-31)
Akshaya Kumar Swain, Lina Pradhan, Dhirendra Bahadur
ABSTRACT
In light of the growing interest in the search for cheap and effective solutions for cancer treatment, we report a simple one pot synthesis of polymer stabilized iron oxide-graphene (PIG) that could be realized on a large scale. The structural (Fe3O4 particle size of ∼11 nm), functional (various oxygen containing moieties), and magnetic (moment of ∼43 emu/g) properties of PIG are explored using various characterization techniques for possible biomedical applications. PIG shows good colloidal stability and is biocompatible even at higher concentrations (2.5 mg/mL) by virtue of cross-linking polymers. The biocompatibility of the composite has been tested using HeLa cell lines by computing the percentage of the reactive oxygen species through the 2,7-dichlorofluorescein (DCF) intensity level. PIG has the ability to load and release both hydrophobic and hydrophilic drugs with a good loading efficiency and capacity. The dug loading efficiency of PIG is measured to be ∼87% and ∼91% for doxorubicin (DOX) and paclitaxel (PTXL), respectively. Under an AC magnetic field, superparamagnetic PIG (2.5 mg/mL) takes less than 16 min to reach the stable hyperthermia temperature, suggesting it as a good anticancer material. A time-dependent cellular uptake of doxorubicin-conjugated PIG has been studied to optimize the parameters for thermo-chemotherapy of cancer. The synergetic effect of both the drug and hyperthermia is observed in the killing of the cancerous cells, verified by computing the cell apoptotic population using a flow cytometer. However, it has been noticed that, even in the absence of chemotherapy, PIG shows good antiproliferative activity with thermotherapy alone.
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard