  • Search Results
  • Universal conventional and real-time PCR diagnosis tools for Sarcoptes scabiei.
Universal conventional and real-time PCR diagnosis tools for Sarcoptes scabiei.
Parasites & vectors (2015-11-17)
Samer Angelone-Alasaad, AnnaRita Molinar Min, Mario Pasquetti, Abdulaziz N Alagaili, Stefano D'Amelio, Federica Berrilli, Vincent Obanda, Mohamed A Gebely, Ramón C Soriguer, Luca Rossi
ABSTRACT
The mite Sarcoptes scabiei has a known host-range of over 100 mammal species including humans. One of the prime objectives of the Sarcoptes-World Molecular Network (WMN) is to design and develop universal Sarcoptes PCR-based diagnosis methods. We describe here for the first time two universal mitochondrial-based diagnosis methods: (i) conventional end-point PCR and (ii) TaqMan real-time PCR. The design of both of these universal diagnosis methods was based on Sarcoptes samples collected from 23 host species in 14 countries. These methods, based on skin scrapings, were successfully used to etiologically confirm the diagnosis of different clinical degrees of sarcoptic mange in 48 animals belonging to six species. These universal PCR-based diagnosis methods are highly specific, technically sensitive and simple, and are based on the amplification of 135 bp from the Mitochondrial 16S rDNA. The method based on TaqMan real-time qPCR was more sensitive than the conventional end-point PCR. Two universal PCR-based diagnosis methods for S. scabiei were successfully designed and applied; one based on conventional end-point PCR and the other on TaqMan real-time PCR. We recommend further testing and the application of these new universal methods worldwide.
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR