  • Search Results
  • Effect of resveratrol and lipoic acid on sirtuin-regulated expression of metabolic genes in bovine liver and muscle slice cultures.
Effect of resveratrol and lipoic acid on sirtuin-regulated expression of metabolic genes in bovine liver and muscle slice cultures.
Journal of animal science (2015-10-07)
Y Ghinis-Hozumi, L González-Dávalos, A Antaramian, F Villarroya, E Piña, A Shimada, A Varela-Echavarría, O Mora
ABSTRACT
Sirtuins (Sirt) are NAD-dependent deacetylases that are activated by the antioxidants resveratrol (RSV) and lipoic acid (LA). The objective of this study was to determine in bovine liver and muscle slice cultures the effect of RSV and LA treatment on the expresssion of Sirt1, Sirt3, peroxisome proliferator-activated receptor γ coactivator 1α (PPARGC1A), and the forkhead box O transcription factors FoxO1 and FoxO3 as well as other factors involved in glucose and lipid metabolism and related to Sirt activity. Tissue slices from crossbred bulls were treated during 60 min with 40 or 80 μ RSV and 30, 100, 300, or 1,000 μ LA under restricted conditions (Krebs-Ringer buffer without nutrients) and fed conditions (2.5 m propionate in combination with 1 n glucagon) for liver slices or with 0.01 μ epinephrine for muscle slices. Quantitative real-time PCR was used to analyze the expression of the mRNA for the genes studied and western blot analysis for the expression of the protein for Sirt1. Our results show that the expression of the mRNA for Sirt1 was enhanced by RSV in liver under restriction ( ≤ 0.0112) and by LA in muscle, more under restriction ( ≤ 0.0121) than after epinephrine administration ( < 0.0001). Sirt3 is affected in a dose-dependent manner by both compounds in both tissues and under both metabolic conditions ( ≤ 0.0452). The expression of the protein for Sirt1 was increased by LA in both tissues under restricted conditions ( = 0.0026 and = 0.0201, respectively) but in liver also in fed conditions ( = 0.0016). Genes involved in the antioxidant response were upregulated in both tissues. These results indicate that bovine Sirt respond differently to RSV and LA stimulation than monogastric Sirt do and that gluconeogenesis in ruminants is not related to Sirt to the same degree as in monogastric species. However, these results provide information about the possible role of Sirt in ruminant metabolism.
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%