  • Search Results
  • Seminal fluid factors regulate activin A and follistatin synthesis in female cervical epithelial cells.
Seminal fluid factors regulate activin A and follistatin synthesis in female cervical epithelial cells.
Molecular and cellular endocrinology (2015-09-30)
David J Sharkey, John E Schjenken, David G Mottershead, Sarah A Robertson
ABSTRACT
Seminal fluid induces pro-inflammatory cytokines and elicits an inflammation-like response in the cervix. Here, Affymetrix microarray and qPCR was utilised to identify activin A (INHBA) and its inhibitor follistatin (FST) amongst the cytokines induced by seminal plasma in Ect1 ectocervical epithelial cells, and a similar response was confirmed in primary ectocervical epithelial cells. TGFB is abundant in seminal plasma and all three TGFB isoforms induced INHBA in Ect1 and primary cells, and neutralisation of TGFB in seminal plasma suppressed the INHBA response. Bacterial lipopolysaccharide in seminal plasma also elicited INHBA, but potently suppressed FST production. There was moderate reciprocal inhibition between FST and INHBA, and cross-attenuating effects were seen. These data identify TGFB and potentially LPS as factors mediating seminal plasma-induced INHBA synthesis in cervical cells. INHBA and FST induced by seminal fluid in cervical tissues may thus contribute to regulation of the post-coital response in women.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture