  • Search Results
  • Low-glucose enhances keratocyte-characteristic phenotype from corneal stromal cells in serum-free conditions.
Low-glucose enhances keratocyte-characteristic phenotype from corneal stromal cells in serum-free conditions.
Scientific reports (2015-06-04)
James W Foster, Ricardo M Gouveia, Che J Connon
ABSTRACT
The avascular cornea is a uniquely-isolated organ, with its stroma constituting a nutrient-poor environment. Consequently, the availability of metabolites such as glucose to corneal stromal cells is considerably reduced compared with other tissues, or indeed with media commonly used to culture these cells in vitro. However, the role of glucose in the behaviour of human corneal keratocytes has been overlooked. As such, we sought to investigate the effects of low-glucose formulations on the phenotype of human corneal stromal cells. Cells cultured in low-glucose were able to survive for extended periods when compared to high-glucose, serum-free conditions. Furthermore, low-glucose enhanced their reversal to a keratocyte-characteristic phenotype. Specifically, cells within low-glucose medium assumed dendritic morphologies, with bean-shaped condensed nuclei, absence of alpha-smooth muscle actin or stress fibres, and a corresponding reduction in migratory and contractile activities when compared with high-glucose, serum-free conditions. Moreover, cells within low-glucose uniquely recovered the ability to express a robust keratocyte-characteristic marker, CD34, while still expressing elevated levels of other representative phenotypic markers such as keratocan, lumican, ALDH1A1, and ALDH3A1. These results indicate that low-glucose enhances keratocyte-characteristic phenotype above and beyond established media formulations and thus has important implications for corneal biology in health and disease.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%