  • Search Results
  • Purification and partial characterization of a thermostable antimicrobial protein from Bacillus subtilis FB123.
Purification and partial characterization of a thermostable antimicrobial protein from Bacillus subtilis FB123.
World journal of microbiology & biotechnology (2015-05-20)
Bihong Shi, Hong Zheng, Jianzhong Huang, Xiuzhen Luo, Xiaolei Luo
ABSTRACT
Antimicrobial proteins/peptides have attracted much attention because of their potential use in the industrial setting. In the present study, a thermostable antimicrobial protein (BSAMP) was purified from the culture supernatant of Bacillus subtilis FB123 by ammonium sulfate precipitation, gel chromatography on Sephacryl S-200 High Resolution, and ion exchange chromatography on DEAE Sepharose Fast Flow column. The molecular weight of the purified BSAMP was 54 kDa, as estimated by sodium dodecyl sulfate-polyacrylamide gel electrophoresis both in the absence and presence of β-mercaptoethanol. Its isoelectric point was determined to be 5.24 by isoelectric focusing electrophoresis. Periodic acid-Schiff staining revealed BSAMP to be a glycoprotein. Maximum activity was obtained at pH 6.0, with over 79% maximum activity retained at pH 3.0-5.0 and pH 7.0-9.0, respectively. BSAMP was shown to be highly thermostable, as its activity did not change obviously after treatment at 100 °C. However, it was partially sensitive to papain, trypsin, and alkali proteases. Finally, the bacterial protein exhibited broad-spectrum antimicrobial activity against several pathogenic organisms. These findings suggested that BSAMP should be further developed as a natural antibacterial agent for disease prevention in aquiculture and agriculture.
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Kanamycin sulfate Streptomyces kanamyceticus, powder, γ-irradiated
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M