  • Search Results
  • miR-145 suppress the androgen receptor in prostate cancer cells and correlates to prostate cancer prognosis.
miR-145 suppress the androgen receptor in prostate cancer cells and correlates to prostate cancer prognosis.
Carcinogenesis (2015-05-15)
Olivia Larne, Zandra Hagman, Hans Lilja, Anders Bjartell, Anders Edsjö, Yvonne Ceder
ABSTRACT
Androgen signalling through the androgen receptor (AR) is essential for prostate cancer initiation, progression and transformation to the lethal castration-resistant state. The aim of this study was to characterize the mechanisms by which miR-145 deregulation contribute to prostate cancer progression. The miR-145 levels, measured by quantitative reverse transcription-polymerase chain reaction, were found to inversely correlate with occurrence of metastases, survival and androgen deprivation therapy response in a well-characterized prostate cancer cohort. Introduction of ectopic miR-145 in prostate cancer cells generated an inhibitory effect on the AR at both transcript and protein levels as well as its activity and downstream targets prostate-specific antigen (PSA), kallikrein-related peptidase 2 and TMPRSS2. The regulation was shown to be mediated by direct binding using Ago2-specific immunoprecipitation, but there was also indication of synergetic AR activation. These findings were verified in clinical prostate specimens by demonstrating inverse correlations between miR-145 and AR expression as well as serum PSA levels. In addition, miR-145 was found to regulate androgen-dependent cell growth in vitro. Our findings put forward novel possibilities of therapeutic intervention, as miR-145 potentially could decrease both the stem cells and the AR expressing bulk of the tumour and hence reduce the transformation to the deadly castration-resistant form of prostate cancer.
Product Description
SAFC
Tromethamine
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
p-Toluidine, 99.6%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
p-Toluidine, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%