  • Search Results
  • Persistent reduced oxygen requirement following blood transfusion during recovery from hemorrhagic shock.
Persistent reduced oxygen requirement following blood transfusion during recovery from hemorrhagic shock.
Respiratory physiology & neurobiology (2015-04-26)
Philippe Haouzi, Andry Van de Louw
ABSTRACT
Our study intended to determine the effects on oxygen uptake (VO2) of restoring a normal rate of O2 delivery following blood transfusion (BT) after a severe hemorrhage (H). Spontaneously breathing urethane anesthetized rats were bled by removing 20 ml/kg of blood over 30 min. Rats were then infused with their own shed blood 15 min after the end of H. At mid-perfusion, half of the rats received a unique infusion of the decoupling agent 2,4-dinitrophenol (DNP, 6 mg/kg). VO2 and arterial blood pressure (ABP) were continuously measured throughout the study, along with serial determination of blood lactate concentration [La]. Animals were euthanized 45 min after the end of reperfusion; liver and lungs were further analyzed for early expression of oxidative stress gene using RT-PCR. Our bleeding protocol induced a significant decrease in ABP and increase in [La], while VO2 dropped by half. The O2 deficit progressively accumulated during the period of bleeding reached -114 ± 53 ml/kg, just before blood transfusion. Despite the transfusion of blood, a significant O2 deficit persisted (-82 ± 59 ml/kg) 45 min after reperfusion. This slow recovery of VO2 was sped up by DNP injection, leading to a fast recovery of O2 deficit after reperfusion, becoming positive (+460 ± 132 ml/kg) by the end of the protocol, supporting the view that O2 supply is not the main controller of VO2 dynamics after BT. Of note is that DNP also enhanced oxidative stress gene expression (up-regulation of NADPH oxidase 4 in the lung for instance). The mechanism of slow recovery of O2 requirement/demand following BT and the resulting effects on tissues exposed to relatively high O2 partial pressure are discussed.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
3-Benzidino-6-(4-chlorophenyl)pyridazine, ≥98% (HPLC), solid
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
1-Bromo-3-chloropropane, 99%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
1-Bromo-3-chloropropane, for isolation of RNA