  • Search Results
  • MicroRNA involvement in a metastatic non-functioning pituitary carcinoma.
MicroRNA involvement in a metastatic non-functioning pituitary carcinoma.
Pituitary (2015-04-12)
Zhenqing Wei, Cuiqi Zhou, Mei Liu, Yong Yao, Jian Sun, Jianqi Xiao, Wenbin Ma, Huijuan Zhu, Renzhi Wang
ABSTRACT
Pituitary carcinomas are extremely rare neoplasms, and molecular events leading to malignant pituitary transformation are largely unknown. Enhanced understanding of molecular mechanisms driving malignant pituitary progression would be beneficial for pituitary carcinoma diagnosis and treatment. Differential microRNA expression in paired primary and metastatic pituitary carcinoma specimens were detected using high-throughput human microRNA microarrays and TaqMan microRNA arrays. Three of significantly deregulated miRNAs were further confirmed using quantitative real-time PCR in the metastatic carcinoma, six atypical pituitary adenomas and eight typical pituitary adenomas. Target genes of microRNAs were bioinformatically predicated and verified in vitro by Western blotting and real-time PCR and in vivo by immunohistochemistry respectively. We present a case of a 50-year-old woman harboring non-functioning pituitary carcinoma with multiple intracranial metastases, and identified up-regulation of miR-20a, miR-106b and miR-17-5p in the metastatic carcinoma as compared to the primary neoplasm. Furthermore, miR-20a and miR-17-5p were increased in the metastatic carcinoma and six atypical pituitary adenomas as compared to eight typical pituitary adenomas as measured by quantitative real-time PCR. Both PTEN and TIMP2 were bioinformatically predicated and confirmed in vitro as target genes of these three microRNAs. As semi-quantified by immunohistochemistry, PTEN was absent and TIMP2 was decreased in the metastatic pituitary carcinoma as compared to pituitary adenomas. Our results suggest microRNA involvement in malignant pituitary progression, whereby increased miR-20a, miR-106b and miR-17-5p promote metastasis by attenuating PTEN and TIMP2 in pituitary carcinoma.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O