MilliporeSigma
Quantitative imaging of light-triggered doxorubicin release.
Biomedical optics express (2015-09-30)
Jeremy Kress, Daniel J Rohrbach, Kevin A Carter, Dandan Luo, Shuai Shao, Shashikant Lele, Jonathan F Lovell, Ulas Sunar
ABSTRACT
The efficacy of chemotherapy is related, in large part, to the concentration of drug that reaches tumor sites. Doxorubicin (DOX) is a common anti-cancer drug that is also approved for use in liposomal form for the treatment of ovarian cancer. We recently developed a porphyrin-phospholipid (PoP)-liposome system that enables on demand release of DOX from liposomes using near infrared irradiation to improve DOX bioavailability. Owing to its intrinsic fluorescence, it is possible, and desirable, to quantify DOX concentration and distribution, preferably noninvasively. Here we quantified DOX distribution following light-triggered drug release in phantoms and an animal carcass using spatial frequency domain imaging. This study demonstrates the feasibility of non-invasive quantitative mapping of DOX distributions in target areas.
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)