MilliporeSigma
Microfluidic Crystal Engineering of π-Conjugated Polymers.
ACS nano (2015-07-17)
Gang Wang, Nils Persson, Ping-Hsun Chu, Nabil Kleinhenz, Boyi Fu, Mincheol Chang, Nabankur Deb, Yimin Mao, Hongzhi Wang, Martha A Grover, Elsa Reichmanis
ABSTRACT
Very few studies have reported oriented crystallization of conjugated polymers directly in solution. Here, solution crystallization of conjugated polymers in a microfluidic system is found to produce tightly π-stacked fibers with commensurate improved charge transport characteristics. For poly(3-hexylthiophene) (P3HT) films, processing under flow caused exciton bandwidth to decrease from 140 to 25 meV, π-π stacking distance to decrease from 3.93 to 3.72 Å and hole mobility to increase from an average of 0.013 to 0.16 cm(2) V(-1) s(-1), vs films spin-coated from pristine, untreated solutions. Variation of the flow rate affected thin-film structure and properties, with an intermediate flow rate of 0.25 m s(-1) yielding the optimal π-π stacking distance and mobility. The flow process included sequential cooling followed by low-dose ultraviolet irradiation that promoted growth of conjugated polymer fibers. Image analysis coupled with mechanistic interpretation supports the supposition that "tie chains" provide for charge transport pathways between nanoaggregated structures. The "microfluidic flow enhanced semiconducting polymer crystal engineering" was also successfully applied to a representative electron transport polymer and a nonhalogenated solvent. The process can be applied as a general strategy and is expected to facilitate the fabrication of high-performance electrically active polymer devices.
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
1,2-Dichlorobenzene, anhydrous, 99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1,2-Dichlorobenzene solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Toluene, anhydrous, 99.8%