  • Search Results
  • Development of topical hydrogels of terbinafine hydrochloride and evaluation of their antifungal activity.
Development of topical hydrogels of terbinafine hydrochloride and evaluation of their antifungal activity.
Drug development and industrial pharmacy (2014-03-01)
Nevin Çelebi, Seda Ermiş, Semiha Özkan
ABSTRACT
The purpose of this study was to prepare hydrogels and microemulsion (ME)-based gel formulations containing 1% terbinafine hydrochloride (TER-HCL) and to evaluate the use of these formulations for the antifungal treatment of fungal infections. Three different hydrogel formulations were prepared using chitosan, Carbopol® 974 and Natrosol® 250 polymers. A pseudo-ternary phase diagram was constructed, and starting from ME formulation, a ME gel form containing 1% Carbopol 974 was prepared. We also examined the characteristic properties of the prepared hyrogels. The physical stability of hydrogels and the ME -based gels were evaluated after storage at different temperatures for a period of 3 months. The release of TER-HCL from the gels and the commercial product (Lamisil®) was carried out by using a standard dialysis membrane in phosphate buffer (pH 5.2) at 32 °C. The results of the in vitro release study showed that the Natrosol gel released the highest amount of drug, followed by Carbopol gel, chitosan gel, commercial product, and the microoemulsion-based gel in that order. In vitro examination of antifungal activity revealed that all the prepared and commercial products were effective against Candida parapsilosis, Penicillium, Aspergillus niger and Microsporum. These results indicate that the Natrosol®-based hydrogel is a good candidate for the topical delivery of TER-HCL.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
MOPS
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)