  • Search Results
  • Involvement of Rho-associated protein kinase (ROCK) and bone morphogenetic protein-binding endothelial cell precursor-derived regulator (BMPER) in high glucose-increased alkaline phosphatase expression and activity in human coronary artery smooth muscle cells.
Involvement of Rho-associated protein kinase (ROCK) and bone morphogenetic protein-binding endothelial cell precursor-derived regulator (BMPER) in high glucose-increased alkaline phosphatase expression and activity in human coronary artery smooth muscle cells.
Cardiovascular diabetology (2015-08-13)
Yuya Terao, Seimi Satomi-Kobayashi, Ken-ichi Hirata, Yoshiyuki Rikitake
ABSTRACT
Vascular calcification is an independent risk factor for cardiovascular disease. Diabetes mellitus increases the incidence of vascular calcification; however, detailed molecular mechanisms of vascular calcification in diabetes mellitus remain unknown. We recently reported that bone morphogenetic protein-binding endothelial cell precursor-derived regulator (BMPER) regulates osteoblast-like trans-differentiation of human coronary artery smooth muscle cells (HCASMCs). We investigated the effect of a hydroxymethylglutaryl-coenzyme A reductase inhibitor (statin), commonly used in patients with atherosclerotic diseases and diabetes mellitus, on alkaline phosphatase (ALP) mRNA expression in aortas of streptozotocin-induced diabetic mice. We also investigated the effects of the statin, Rho-associated protein kinase (ROCK) inhibitors and BMPER knockdown on ALP mRNA expression and activity in HCASMCs cultured in high glucose-containing media. Alkaline phosphatase mRNA expression was increased in aortas of streptozotocin-induced diabetic mice, and the increase was inhibited by rosuvastatin. ALP mRNA expression and activity were increased in HCASMCs cultured in high glucose-containing media, and the increases were suppressed by rosuvastatin. This suppression was reversed by the addition of mevalonate or geranylgeranyl pyrophosphate, but not farnesyl pyrophosphate. High glucose-increased ALP mRNA expression and activity were suppressed by ROCK inhibitors. Moreover, BMPER mRNA expression was increased in diabetic mouse aortas and in HCASMCs cultured in high glucose-containing media, but was not inhibited by rosuvastatin or ROCK inhibitors. Knockdown of BMPER suppressed high glucose-increased ALP activity, but not ROCK activity in HCASMCs. There are at least two independent pathways in high glucose-induced ALP activation in HCASMCs: the Rho-ROCK signaling pathway and the BMPER-dependent pathway.
Sigma-Aldrich
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
Sigma-Aldrich
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR