  • Search Results
  • Multiple shape memory polymers based on laminates formed from thiol-click chemistry based polymerizations.
Multiple shape memory polymers based on laminates formed from thiol-click chemistry based polymerizations.
Soft matter (2015-08-04)
M Podgórski, C Wang, C N Bowman
ABSTRACT
This investigation details the formation of polymer network trilayer laminates formed by thiol-X click chemistries, and their subsequent implementation and evaluation for quadruple shape memory behavior. Thiol-Michael addition and thiol-isocyanate-based crosslinking reactions were employed to fabricate each of the laminate's layers with independent control of the chemistry and properties of each layer and outstanding interlayer adhesion and stability. The characteristic features of step-growth thiol-X reactions, such as excellent network uniformity and narrow thermal transitions as well as their stoichiometric nature, enabled fabrication of trilayer laminates with three distinctly different glass transition temperatures grouped within a narrow range of 100 °C. Through variations in the layer thicknesses, a step-wise modulus drop as a function of temperature was achieved. This behavior allowed multi-step programming and the demonstration and quantification of quadruple shape memory performance. As is critical for this performance, the interface connecting the layers was evaluated in stoichiometric as well as off-stoichiometric systems. It was shown that the laminated structures exhibit strong interfacial binding and hardly suffer any delamination during cyclic material testing and deformation.
Sigma-Aldrich
Trimethylolpropane triacrylate, contains 600 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Sigma-Aldrich
Toluene 2,4-diisocyanate, purum, suitable for electron microscopy, ≥98.0% (GC)
Sigma-Aldrich
Tolylene-2,4-diisocyanate, 95%
Sigma-Aldrich
Pentaerythritol, 98%
Sigma-Aldrich
Pentaerythritol, 99%
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1.6 mmol/g loading
Sigma-Aldrich
Rhodamine B, for fluorescence
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3 mmol/g triphenylphosphine loading
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Tolylene-2,4-diisocyanate, technical grade, 80%
Sigma-Aldrich
4,4′-Methylenebis(cyclohexyl isocyanate), mixture of isomers, 90%
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.2 mmol/g loading
Sigma-Aldrich
Divinyl sulfone, contains hydroquinone as inhibitor, ≥96%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Hexamethylene diisocyanate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)