MilliporeSigma
  • Strategies for Coordination of a Serosurvey in Parallel with an Immunization Coverage Survey.
Strategies for Coordination of a Serosurvey in Parallel with an Immunization Coverage Survey.
The American journal of tropical medicine and hygiene (2015-06-10)
Mark A Travassos, Berhane Beyene, Zenaw Adam, James D Campbell, Nigisti Mulholland, Seydou S Diarra, Tassew Kassa, Lisa Oot, Jenny Sequeira, Mardi Reymann, William C Blackwelder, Marcela F Pasetti, Samba O Sow, Robert Steinglass, Amha Kebede, Myron M Levine
ABSTRACT
A community-based immunization coverage survey is the standard way to estimate effective vaccination delivery to a target population in a region. Accompanying serosurveys can provide objective measures of protective immunity against vaccine-preventable diseases but pose considerable challenges with respect to specimen collection and preservation and community compliance. We performed serosurveys coupled to immunization coverage surveys in three administrative districts (woredas) in rural Ethiopia. Critical to the success of this effort were serosurvey equipment and supplies, team composition, and tight coordination with the coverage survey. Application of these techniques to future studies may foster more widespread use of serosurveys to derive more objective assessments of vaccine-derived seroprotection and monitor and compare the performance of immunization services in different districts of a country.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles