  • Search Results
  • GC/MS method for determining carbon isotope enrichment and concentration of underivatized short-chain fatty acids by direct aqueous solution injection of biogas digester samples.
GC/MS method for determining carbon isotope enrichment and concentration of underivatized short-chain fatty acids by direct aqueous solution injection of biogas digester samples.
Talanta (2015-06-17)
Daniel Girma Mulat, Anders Feilberg
ABSTRACT
In anaerobic digestion of organic matter, several metabolic pathways are involved during the simultaneous production and consumption of short-chain fatty acids (SCFA) in general and acetate in particular. Understanding the role of each pathway requires both the determination of the concentration and isotope enrichment of intermediates in conjunction with isotope labeled substrates. The objective of this study was to establish a rapid and simple GC/MS method for determining the isotope enrichment of acetate and concentration of underivatized short-chain fatty acids (SCFA) in biogas digester samples by direct liquid injection of acidified aqueous samples. Sample preparation involves only acidification, centrifugation and filtration of the aqueous solution followed by direct injection of the aqueous supernatant solution onto a polar column. With the sample preparation and GC/MS conditions employed, well-resolved and sharp peaks of underivatized SCFA were obtained in a reasonably short time. Good recovery (96.6-102.3%) as well as low detection (4-7 µmol/L) and quantification limits (14-22 µmol/L) were obtained for all the 6 SCFA studied. Good linearity was achieved for both concentration and isotope enrichment measurement with regression coefficients higher than 0.9978 and 0.9996, respectively. The method has a good intra- and inter-day precision with a relative standard deviation (RSD) below 6% for determining the tracer-to-tracee ratio (TTR) of both [2-(13)C]acetate and [U-(13)C]acetate. It has also a good intra- and inter-day precision with a RSD below 6% and 5% for determining the concentration of standard solution and biogas digester samples, respectively. Acidification of biogas digester samples with oxalic acid provided the low pH required for the protonation of SCFA and thus, allows the extraction of SCFA from the complex sample matrix. Moreover, oxalic acid was the source of formic acid which was produced in the injector set at high temperature. The produced formic acid prevented the adsorption of SCFA in the column, thereby eliminating peak tailing and ghost peaks. The applicability of the validated GC/MS method for determining the concentration of acetate and its (13)C isotope enrichment in anaerobic digester samples was tested and the results demonstrated the suitability of this method for identifying the metabolic pathways involved in degradation and production of acetate.
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG
SAFC
Acetic acid, glacial
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Isovaleric acid, ≥99%, FCC, FG
Sigma-Aldrich
Isovaleric acid, 99%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Valeric acid, ≥99%, FCC, FG
Sigma-Aldrich
Isobutyric acid, ≥99%, FCC, FG
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%