  • Search Results
  • Inhibition of cellular transdifferentiation by losartan minimizes but does not reverse type 2 diabetes-induced renal fibrosis.
Inhibition of cellular transdifferentiation by losartan minimizes but does not reverse type 2 diabetes-induced renal fibrosis.
Journal of the renin-angiotensin-aldosterone system : JRAAS (2014-03-05)
Carine P Arnoni, Edgar Maquigussa, Clévia S Passos, Luciana G Pereira, Mirian A Boim
ABSTRACT
Transformer Growth Factor (TGF-β1) and angiotensin II (AngII) induce epithelial mesenchymal transition (EMT) and myofibroblastic transdifferentiation (MFT) contributing to renal fibrosis. The present study evaluated the capacity of an AT1 receptor blocker (losartan) to induce the regression of pre-existing fibrosis via interference with MFT and EMT in a rat model of type 2 diabetes, and in cultured mesangial cells (MCs) stimulated with high glucose and AngII. After 12 weeks of diabetes induction (D12 group), animals showing evidence of nephropathy, were divided in groups untreated for additional 8 weeks (D20 group) and treated for additional 8 weeks with losartan (D20+los group). D12 animals presented hyperglycemia, insulin resistance, hypertension, proteinuria, increased levels of TGF-β1 and MFT/EMT markers. Losartan stabilized all of these parameters and hindered the progression of fibrosis, but it did not reverse the pre-existing fibrotic manifestations. Losartan reduced TGF-β1 in the tubules, but not in the glomeruli. Stimulated MC exhibited myofibroblast phenotype and capacity for migration, which were completely reversed by losartan. Cellular transition may play a role in diabetes-inducing renal fibrogenesis in both AngII-TGF-β1 axis-dependent and independent manners. Losartan was efficient in preventing cells from undergoing further transdifferentiation, but this strategy was not sufficient to induce regression of the pre-existing tissue fibrosis.
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O